Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136241.1 | NP_001129713.1 | 1623 | |||
GenBank mRNA | BC078722.1 | - | 5 | 619 | ||
BC091122.1 | - | 765 |