Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014906.4 | NP_055721.3 | 7 | 5037 | ||
NR_048561.1 | 6 | 4837 | ||||
XM_005257169.4 | XP_005257226.1 | 6 | 4262 | |||
XM_011524534.2 | XP_011522836.1 | 6 | 5158 | |||
XM_017024359.1 | XP_016879848.1 | 7 | 5554 | |||
GenBank mRNA | AF260269.1 | - | 7 | 5067 |