Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139084.2 | NP_620784.2 | 2 | 511 | ||
XM_006233058.2 | XP_006233120.1 | 2 | 499 | |||
XM_008761367.1 | XP_008759589.1 | 2 | 499 | |||
GenBank mRNA | AF255614.1 | - | 2 | 520 |