Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139037.1 | NP_620606.1 | 3 | 358 | ||
XM_006228381.2 | XP_006228443.1 | 4 | 522 | |||
GenBank mRNA | AF347056.1 | - | 3 | 358 |
Gene Symbol: LOC100911185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759080.1 | XP_008757302.1 | 4 | 475 |