Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPN20 SIRNA Assays ›
Gene Symbol:
PTPN20
Gene Name:
protein tyrosine phosphatase, non-receptor type 20
Gene Aliases:
CT126, PTPN20A, PTPN20B, bA142I17.1, bA42B19.1
Chromosome Location:
Chr.10: 46911134 - 47003834 on Build GRCh38
UniGene ID:
Hs.440733
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
5818
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPN20
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042357.4
NP_001035816.1
6, 7
720
NM_001042358.4
NP_001035817.1
5, 6
578
NM_001042361.4
NP_001035820.1
5, 6
578
NM_001042363.4
NP_001035822.1
5, 6
563
NM_001042364.4
NP_001035823.1
4, 5
468
NM_001320681.1
NP_001307610.1
4, 5
468
NM_001320683.1
NP_001307612.1
6, 7
735
NM_001320685.1
NP_001307614.1
5, 6
640
NM_001320686.1
NP_001307615.1
4, 5
483
NM_015605.8
NP_056420.3
6, 7
720
XM_011539605.2
XP_011537907.1
6, 7
864
XM_011539606.2
XP_011537908.1
6, 7
1956
XM_011539607.2
XP_011537909.1
6, 7
855
XM_011539610.2
XP_011537912.1
3, 4
338
XM_011539611.2
XP_011537913.1
1, 2
21
XM_017016042.1
XP_016871531.1
6, 7
699
XM_017016043.1
XP_016871532.1
6, 7
699
XM_017016044.1
XP_016871533.1
5, 6
621
XM_017016046.1
XP_016871535.1
6, 7
699
XM_017016047.1
XP_016871536.1
6, 7
699
XM_017016048.1
XP_016871537.1
6, 7
699
XM_017016049.1
XP_016871538.1
4, 5
430
XM_017016050.1
XP_016871539.1
3, 4
632
XM_017016051.1
XP_016871540.1
6, 7
699
XM_017016052.1
XP_016871541.1
6, 7
699
XM_017016054.1
XP_016871543.1
5, 6
621
XM_017016059.1
XP_016871548.1
4, 5
610
GenBank mRNA
AK309922.1
-
5, 6
782
AL050040.1
-
4, 5
426
AY704141.1
-
5, 6
519
AY704144.1
-
5, 6
519
AY704146.1
-
5, 6
568
AY704147.1
-
6, 7
676
AY704150.1
-
5, 6
581
AY704151.1
-
4, 5
473
AY704152.1
-
4, 5
424
AY704154.1
-
4, 5
424
BC093750.1
-
5, 6
574
7939595
BC113473.1
-
5, 6
574
8322525
Back To Top
More Information
Panther Classification:
Molecular Function -
protein phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0