Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322227.1 | NP_001309156.1 | 3 | 492 | ||
NM_001322228.1 | NP_001309157.1 | 2 | 352 | |||
NM_001322229.1 | NP_001309158.1 | 2 | 352 | |||
NM_001322230.1 | NP_001309159.1 | 2 | 352 | |||
NM_001322231.1 | NP_001309160.1 | 2 | 352 | |||
NM_001322232.1 | NP_001309161.1 | 2 | 352 | |||
NM_001322233.1 | NP_001309162.1 | 2 | 352 | |||
NM_001322234.1 | NP_001309163.1 | 2 | 352 | |||
NM_001322235.1 | NP_001309164.1 | 3 | 492 | |||
NM_001322236.1 | NP_001309165.1 | 2 | 352 | |||
NM_001322237.1 | NP_001309166.1 | 2 | 151 | |||
NM_004755.3 | NP_004746.2 | 2 | 352 | |||
NM_182398.2 | NP_872198.1 | 2 | 352 | |||
XM_017021786.1 | XP_016877275.1 | 2 | 323 | |||
GenBank mRNA | AF074393.1 | - | 2 | 302 | ||
AF080000.1 | - | 2 | 318 | |||
AF090421.1 | - | 2 | 301 | |||
AK295266.1 | - | 2 | 151 | |||
AK309925.1 | - | 2 | 339 | |||
BC017187.2 | - | 2 | 303 | 3139114 | ||
BG699153.1 | - | 2 | 387 |