Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145671.1 | NP_663704.1 | 5 | 824 | ||
XM_006239521.2 | XP_006239583.1 | 5 | 851 | |||
XM_006239526.2 | XP_006239588.1 | 5 | 989 | |||
XM_008764336.1 | XP_008762558.1 | 5 | 851 | |||
GenBank mRNA | AF505655.1 | - | 5 | 824 |