Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BFAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016561.2 | NP_057645.1 | 8 | 2096 | ||
XM_005255350.2 | XP_005255407.1 | 7 | 1871 | |||
XM_011522520.1 | XP_011520822.1 | 6 | 1701 | |||
GenBank mRNA | AF173003.1 | - | 8 | 1968 | ||
BC003054.1 | - | 8 | 1927 | 3537783 |