Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153315.2 | NP_695227.2 | 2 | 420 | ||
GenBank mRNA | AB062758.1 | - | 2 | 236 | ||
BC070961.1 | - | 2 | 420 | 7099044 |