Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Decr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_171996.3 | NP_741993.1 | 8 | 770 | ||
XM_006246024.2 | XP_006246086.1 | 9 | 970 | |||
GenBank mRNA | AF044574.1 | - | 8 | 692 | ||
BC070959.1 | - | 8 | 771 | 7097883 |
Gene Symbol: Rab11fip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY383698.1 | - | 815 |