Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhfpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181085.2 | NP_851599.1 | 2 | 547 | ||
XM_008773405.1 | XP_008771627.1 | 3 | 580 | |||
GenBank mRNA | AY253667.1 | - | 2 | 547 |