Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A3R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130012.2 | NP_001123484.1 | 1, 2 | 359 | ||
NM_004785.5 | NP_004776.3 | 1, 2 | 359 | |||
GenBank mRNA | AF004900.1 | - | 1, 2 | 237 | ||
AF035771.1 | - | 1, 2 | 221 | |||
BC014513.1 | - | 1, 2 | 332 | |||
BC069014.1 | - | 1, 2 | 328 | |||
BC106001.1 | - | 1, 2 | 335 | 5296143 | ||
DB495688.2 | - | 1, 2 | 340 | |||
U82108.1 | - | 1, 2 | 312 | |||
Z50150.1 | - | 1, 2 | 314 |