Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001279349.1 | NP_001266278.1 | 1 | 367 | ||
NM_001279350.1 | NP_001266279.1 | 1 | 367 | |||
NM_020385.3 | NP_065118.2 | 1 | 367 | |||
NR_103995.1 | 1 | 367 | ||||
NR_103996.1 | 1 | 367 | ||||
GenBank mRNA | AF273304.1 | - | 1 | 315 | ||
AF295774.1 | - | 1 | 315 | |||
AK025493.1 | - | 1 | 221 | |||
AK304547.1 | - | 1 | 283 | |||
AK310164.1 | - | 1 | 270 | |||
AK314333.1 | - | 1 | 281 | |||
AK315904.1 | - | 2 | 218 | |||
AL136894.1 | - | 1 | 298 | |||
BC009274.2 | - | 1 | 270 | 3958331 | ||
BC071691.1 | - | 1 | 226 | |||
BX333016.2 | - | 1 | 257 | |||
DC387839.1 | - | 1 | 367 |