Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Id1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012797.2 | NP_036929.2 | 1 | 312 | ||
XM_006235267.2 | XP_006235329.1 | 1 | 1278 | |||
GenBank mRNA | BC127448.1 | - | 1 | 324 | 8372348 | |
D10862.1 | - | 1 | 293 | |||
L23148.1 | - | 1 | 313 | |||
M86708.1 | - | 1 | 319 |