Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pemt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013003.1 | NP_037135.1 | 3 | 273 | ||
GenBank mRNA | BC091162.1 | - | 3 | 313 | 7325819 | |
L14441.1 | - | 3 | 273 |