Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eci1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017306.4 | NP_059002.2 | 3 | 173 | ||
GenBank mRNA | BC078705.1 | - | 3 | 173 | 7129389 | |
FQ213384.1 | - | 3 | 181 | |||
FQ218351.1 | - | 3 | 174 | |||
M61112.1 | - | 3 | 162 | |||
X61184.1 | - | 3 | 184 |