Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP3CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142353.2 | NP_001135825.1 | 2 | 295 | ||
NM_001142354.2 | NP_001135826.1 | 2 | 295 | |||
NM_001289968.1 | NP_001276897.1 | 2 | 295 | |||
NM_001289969.1 | NP_001276898.1 | 3 | 414 | |||
NM_021132.3 | NP_066955.1 | 2 | 295 | |||
XM_005269944.1 | XP_005270001.1 | 2 | 295 | |||
XM_017016379.1 | XP_016871868.1 | 2 | 295 | |||
GenBank mRNA | AJ488506.1 | - | 2 | 232 | ||
AK301582.1 | - | 3 | 321 | |||
BC028049.1 | - | 2 | 283 | 4538822 | ||
BC035464.1 | - | 2 | 253 | |||
DA128707.1 | - | 2 | 295 | |||
DA192735.1 | - | 2 | 295 | |||
DA854064.1 | - | 2 | 295 | |||
M29550.1 | - | 2 | 267 | |||
M29551.1 | - | 2 | 276 |