Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syngr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053553.1 | NP_446005.1 | 2 | 359 | ||
GenBank mRNA | AF039085.1 | - | 2 | 359 | ||
FQ218897.1 | - | 202 | ||||
FQ224907.1 | - | 203 |