Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012856.1 | NP_036988.1 | 1 | 1613 | ||
GenBank mRNA | M68880.1 | - | 1 | 1613 | ||
X62840.1 | - | 1 | 738 |