Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019289.2 | NP_062162.1 | 3 | 218 | ||
GenBank mRNA | AF083269.1 | - | 2 | 131 | ||
BC062027.1 | - | 3 | 218 | 5622004 | ||
FQ228791.1 | - | 3 | 189 | |||
FQ228917.1 | - | 3 | 234 | |||
FQ228971.1 | - | 3 | 203 |