Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031019.1 | NP_112281.1 | 2 | 367 | ||
GenBank mRNA | X03036.1 | - | 2 | 365 |