Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkx6-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031737.1 | NP_113925.1 | 2 | 1496 | ||
GenBank mRNA | AF004431.1 | - | 2 | 1496 |