Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181379.2 | NP_852044.1 | 1 | 131 | ||
XM_008760143.1 | XP_008758365.1 | 2 | 702 | |||
GenBank mRNA | AY251278.1 | - | 1 | 84 | ||
BC090324.1 | - | 1 | 131 | 7128996 |