Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019897.2 | NP_063950.2 | 1 | 667 | ||
GenBank mRNA | BC104869.1 | - | 1 | 667 | 8143872 | |
BC113651.1 | - | 1 | 667 | 8322703 |