Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301353.1 | NP_001288282.1 | 4, 5 | 596 | ||
NM_001301354.1 | NP_001288283.1 | 3, 4 | 530 | |||
NM_007470.4 | NP_031496.2 | 4, 5 | 600 | |||
GenBank mRNA | AK135046.1 | - | 4, 5 | 388 | ||
AK157917.1 | - | 4, 5 | 389 | |||
AK157925.1 | - | 4, 5 | 388 | |||
AK158118.1 | - | 4, 5 | 385 | |||
AK158405.1 | - | 4, 5 | 391 | |||
AK160729.1 | - | 3, 4 | 319 | |||
AK162392.1 | - | 4, 5 | 388 | |||
BC145907.1 | - | 2, 3 | 278 | 40131210 | ||
BC145909.1 | - | 2, 3 | 278 | 40131212 | ||
X82648.1 | - | 3, 4 | 334 |