Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177307.1 | NP_001170778.1 | 8 | 992 | ||
NM_001177308.1 | NP_001170779.1 | 7 | 865 | |||
NM_007438.4 | NP_031464.1 | 7 | 915 | |||
GenBank mRNA | AK002561.1 | - | 7 | 805 | ||
AK011238.1 | - | 3 | 325 | |||
AK145639.1 | - | 7 | 802 | |||
AK154295.1 | - | 7 | 798 | |||
AK162290.1 | - | 7 | 801 | |||
BC043026.1 | - | 7 | 791 | 5691975 | ||
BC050896.1 | - | 7 | 833 | 5715436 | ||
BC066218.1 | - | 7 | 811 | 30054842 | ||
BC066801.1 | - | 7 | 811 | 30072521 | ||
BC089495.1 | - | 7 | 844 | 30312668 | ||
EF662059.1 | - | 7 | 846 | |||
EF662060.1 | - | 7 | 796 | |||
X03797.1 | - | 5 | 390 | |||
Y00516.1 | - | 7 | 761 |