Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5ar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173550.1 | NP_001167021.1 | 2 | 727 | ||
NM_007577.4 | NP_031603.2 | 3 | 666 | |||
GenBank mRNA | AK150407.1 | - | 1 | 386 | ||
AK158027.1 | - | 2 | 586 | |||
AY220494.1 | - | 1 | 536 | |||
BC125641.1 | - | 2 | 727 | 40130156 | ||
BC125643.1 | - | 2 | 727 | 40130158 |
Set Membership: |
G-protein coupled receptor |