Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007531.2 | NP_031557.2 | 5 | 675 | ||
GenBank mRNA | AK002334.1 | - | 5 | 641 | ||
AK011107.1 | - | 5 | 642 | |||
AK090088.1 | - | 5 | 631 | |||
AK132052.1 | - | 5 | 637 | |||
AK132058.1 | - | 5 | 664 | |||
AK135729.1 | - | 5 | 593 | |||
AK151952.1 | - | 5 | 669 | |||
AK152375.1 | - | 4 | 1324 | |||
AK153368.1 | - | 4 | 1324 | |||
AK216410.1 | - | 1 | 59 | |||
AK218588.1 | - | 1 | 59 | |||
AK219005.1 | - | 1 | 59 | |||
AY211613.1 | - | 5 | 637 | |||
AY319418.1 | - | 5 | 527 | |||
BC145875.1 | - | 5 | 627 | 40131178 | ||
X78683.1 | - | 5 | 631 |