Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044740.2 | NP_001038205.1 | 12 | 2677 | ||
NM_007514.3 | NP_031540.2 | 12 | 2674 | |||
GenBank mRNA | AK004330.1 | - | 1 | 245 | ||
AK154621.1 | - | 12 | 2651 | |||
BC021653.1 | - | 12 | 2669 | |||
BC127082.1 | - | 12 | 2590 | 40111372 | ||
DQ086834.1 | - | 12 | 2629 | |||
L29006.1 | - | 11 | 2999 | |||
M62838.1 | - | 12 | 2590 |