Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007555.3 | NP_031581.2 | 1 | 894 | ||
GenBank mRNA | AK033362.1 | - | 1 | 897 | ||
BC100751.1 | - | 1 | 165 | |||
BC100752.1 | - | 1 | 165 | |||
BC100753.1 | - | 1 | 165 | |||
BC100754.1 | - | 1 | 165 | |||
BC141283.1 | - | 1 | 697 | 9055895 | ||
L41145.1 | - | 1 | 166 |