Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007621.2 | NP_031647.1 | 3, 4 | 348 | ||
GenBank mRNA | AK081816.1 | - | 3, 4 | 319 | ||
BC010758.1 | - | 3, 4 | 308 | 4219349 | ||
D26123.1 | - | 3, 4 | 335 | |||
X07411.1 | - | 3, 4 | 305 |