Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159555.1 | NP_001153027.1 | 3, 4 | 587 | ||
NM_001159556.1 | NP_001153028.1 | 3, 4 | 447 | |||
NM_001159557.1 | NP_001153029.1 | 4, 5 | 498 | |||
NM_001159558.1 | NP_001153030.1 | 4, 5 | 614 | |||
NM_007643.4 | NP_031669.3 | 3, 4 | 590 | |||
GenBank mRNA | AK052825.1 | - | 3, 4 | 391 | ||
AK128908.1 | - | 4, 5 | 391 | |||
AK150648.1 | - | 3, 4 | 411 | |||
AK151355.1 | - | 3, 4 | 426 | |||
AK152904.1 | - | 1, 2 | 97 | |||
AK172162.1 | - | 4, 5 | 436 | |||
BC010262.1 | - | 3, 4 | 399 | 3481681 | ||
BQ926044.1 | - | 4, 5 | 632 | |||
L23108.1 | - | 3, 4 | 408 |