Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007661.3 | NP_031687.2 | 2 | 162 | ||
GenBank mRNA | AK008283.1 | - | 2 | 139 | ||
AK077668.1 | - | 2 | 163 | |||
AK135402.1 | - | 2 | 169 | |||
AK147133.1 | - | 2 | 140 | |||
AK171785.1 | - | 1 | 721 | |||
BC052920.1 | - | 2 | 121 | 6489832 | ||
L37092.1 | - | 1 | 737 |