Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109761.2 | NP_001103231.1 | 5 | 779 | ||
NM_001177799.1 | NP_001171270.1 | 5 | 1046 | |||
NM_007601.3 | NP_031627.2 | 5 | 1046 | |||
GenBank mRNA | AF091998.1 | - | 5 | 743 | ||
AF127766.2 | - | 5 | 766 | |||
BC090661.1 | - | 5 | 779 | 30681055 | ||
BC139790.1 | - | 5 | 766 | 40047690 | ||
X92523.1 | - | 5 | 766 |