Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007634.4 | NP_031660.3 | 4 | 398 | ||
GenBank mRNA | AK030298.1 | - | 337 | |||
AK031613.1 | - | 4 | 338 | |||
AK032961.1 | - | 4 | 324 | |||
AK082485.1 | - | 4 | 366 | |||
AK085183.1 | - | 4 | 335 | |||
AK159594.1 | - | 4 | 399 | |||
AK162447.1 | - | 4 | 338 | |||
AK169263.1 | - | 4 | 349 | |||
AK211279.1 | - | 3 | 236 | |||
BC021658.1 | - | 4 | 318 | |||
BC037662.1 | - | 4 | 350 | 5353169 | ||
U20612.1 | - | 4 | 336 | |||
U20636.1 | - | 4 | 453 | |||
Z47766.1 | - | 4 | 338 |