Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007650.3 | NP_031676.3 | 3 | 445 | ||
GenBank mRNA | AK042105.1 | - | 3 | 389 | ||
AK089076.1 | - | 3 | 394 | |||
AK143736.1 | - | 3 | 421 | |||
BC011258.1 | - | 3 | 445 | 4188683 | ||
M15177.1 | - | 3 | 323 | |||
U65961.1 | - | 3 | 323 |