Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110320.1 | NP_001103790.1 | 5 | 496 | ||
NM_001110321.1 | NP_001103791.1 | 4 | 424 | |||
NM_001110322.1 | NP_001103792.1 | 3 | 334 | |||
NM_007654.2 | NP_031680.2 | 5 | 496 | |||
GenBank mRNA | AF543213.1 | - | 5 | 397 | ||
AF543214.1 | - | 3 | 172 | |||
AK134132.1 | - | 5 | 499 | |||
AK144191.1 | - | 4 | 427 | |||
AK151094.1 | - | 336 | ||||
BC003824.1 | - | 5 | 454 | 3599554 | ||
BY743956.1 | - | 3 | 334 | |||
J04170.1 | - | 5 | 433 | |||
S40777.1 | - | 5 | 429 |