Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007737.2 | NP_031763.2 | 1, 2 | 457 | ||
GenBank mRNA | AK080811.1 | - | 1, 2 | 239 | ||
AK084285.1 | - | 1, 2 | 239 | |||
AK084803.1 | - | 1, 2 | 94 | |||
AK132413.1 | - | 1, 2 | 239 | |||
AK139130.1 | - | 1, 2 | 32 | |||
AK147220.1 | - | 1, 2 | 239 | |||
AK147328.1 | - | 1, 2 | 458 | |||
AK151929.1 | - | 1, 2 | 241 | |||
AK160008.1 | - | 1, 2 | 31 | |||
BC055077.1 | - | 1, 2 | 208 | 30059991 | ||
L02918.1 | - | 1, 2 | 89 |