Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col11a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007729.2 | NP_031755.2 | 2 | 541 | ||
GenBank mRNA | AK052979.1 | - | 2 | 206 | ||
AK081304.1 | - | 2 | 464 | |||
AK140136.1 | - | 2 | 540 | |||
AK157734.1 | - | 2 | 385 | |||
BC052161.1 | - | 2 | 491 | 6393164 | ||
D38162.1 | - | 2 | 355 |