Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007743.2 | NP_031769.2 | 37 | 2410 | ||
GenBank mRNA | AK075707.1 | - | 15 | 981 | ||
AK132496.1 | - | 37 | 2406 | |||
AK159212.1 | - | 37 | 2411 | |||
AK159408.1 | - | 37 | 2407 | |||
AK159578.1 | - | 37 | 2408 | |||
AK159958.1 | - | 16 | 1029 | |||
AK160098.1 | - | 37 | 2410 | |||
AK160787.1 | - | 37 | 2406 | |||
AK160788.1 | - | 37 | 2405 | |||
AK160790.1 | - | 37 | 2406 | |||
AK160814.1 | - | 29 | 1875 | |||
AK160946.1 | - | 37 | 2409 | |||
AK161017.1 | - | 37 | 2411 | |||
AK161605.1 | - | 37 | 2406 | |||
AK164628.1 | - | 16 | 1029 | |||
AK164808.1 | - | 37 | 2406 | |||
BC007158.1 | - | 37 | 2390 | 3487477 | ||
BC042503.1 | - | 37 | 2398 | 3586272 | ||
X58251.1 | - | 37 | 2346 |