Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007815.3 | NP_031841.3 | 2 | 333 | ||
GenBank mRNA | AI182545.1 | - | 2 | 344 | ||
AK139760.1 | - | 373 | ||||
AK149558.1 | - | 2 | 152 | |||
BC019908.1 | - | 2 | 315 | 5123631 | ||
D17674.1 | - | 2 | 307 |