Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007857.4 | NP_031883.1 | 2 | 687 | ||
GenBank mRNA | AK032466.1 | - | 2 | 670 | ||
AK033392.1 | - | 2 | 688 |