Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Daxx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199733.1 | NP_001186662.1 | 3 | 985 | ||
NM_007829.4 | NP_031855.3 | 3 | 885 | |||
GenBank mRNA | AF006040.1 | - | 2 | 739 | ||
AK090356.1 | - | 3 | 816 | |||
AK133863.1 | - | 3 | 793 | |||
AK144332.1 | - | 3 | 816 | |||
AK145906.1 | - | 3 | 816 | |||
AK146743.1 | - | 3 | 812 | |||
AK151279.1 | - | 3 | 805 | |||
AK153149.1 | - | 3 | 773 | |||
AK156425.1 | - | 3 | 986 | |||
AK172616.1 | - | 3 | 986 | |||
AY734459.1 | - | 1 | 452 | |||
AY734465.1 | - | 1 | 363 | |||
AY734466.1 | - | 499 | ||||
AY758273.1 | - | 1 | 367 | |||
BC128373.1 | - | 3 | 798 | 40111681 | ||
KJ650207.1 | - | 2 | 715 |