Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042623.2 | NP_001036088.1 | 6 | 848 | ||
NM_001271579.1 | NP_001258508.1 | 7 | 975 | |||
NM_007905.3 | NP_031931.2 | 7 | 1004 | |||
GenBank mRNA | AK036370.1 | - | 7 | 964 | ||
AK132756.1 | - | 6 | 806 | |||
AK167590.1 | - | 6 | 837 | |||
BC046535.1 | - | 7 | 985 | 4935713 | ||
BC052394.1 | - | 7 | 1337 | 5719837 | ||
S73882.1 | - | 7 | 960 | |||
S73883.1 | - | 7 | 967 | |||
S73884.1 | - | 7 | 967 | |||
U63386.1 | - | 6 | 926 |