Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021724.4 | NP_068370.1 | 8 | 2564 | ||
GenBank mRNA | AK300155.1 | - | 5 | 1260 | ||
BC047875.1 | - | 8 | 2388 | 6190140 | ||
BC056148.1 | - | 8 | 2455 | 6195844 | ||
M24898.1 | - | 8 | 2560 | |||
X72631.1 | - | 8 | 2135 |
Gene Symbol: THRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M24900.1 | - | 1 | 1299 |