Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007927.3 | NP_031953.1 | 3, 4 | 540 | ||
GenBank mRNA | AK146361.1 | - | 3, 4 | 386 | ||
AK160531.1 | - | 3, 4 | 385 | |||
AK167852.1 | - | 3, 4 | 383 | |||
BC132131.1 | - | 3, 4 | 352 | 40130408 | ||
BC132133.1 | - | 3, 4 | 352 | 40130410 | ||
U73902.1 | - | 3, 4 | 390 |