Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008001.4 | NP_032027.2 | 4 | 1515 | ||
GenBank mRNA | AK148151.1 | - | 4 | 1516 | ||
AK162865.1 | - | 2 | 505 | |||
BC011462.1 | - | 4 | 1452 | 3674293 | ||
U22325.1 | - | 4 | 1114 |