Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146348.1 | NP_001139820.1 | 4 | 756 | ||
NM_001146350.1 | NP_001139822.1 | 4 | 759 | |||
NM_007932.2 | NP_031958.2 | 4 | 759 | |||
GenBank mRNA | AK151302.1 | - | 4 | 729 | ||
AK178235.1 | - | 2 | 59 | |||
AK187426.1 | - | 2 | 59 | |||
AK188120.1 | - | 2 | 59 | |||
AY679531.1 | - | 4 | 759 | |||
BC029080.1 | - | 4 | 673 | 4168250 | ||
S69407.1 | - | 5 | 624 | |||
X77952.1 | - | 4 | 759 |