Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035231.1 | - | 3 | 354 | ||
AK078773.1 | - | 4 | 544 | |||
AK154569.1 | - | 4 | 632 | |||
AK155150.1 | - | 4 | 817 | |||
AK155752.1 | - | 3 | 366 | |||
AK157265.1 | - | 4 | 486 | |||
BC052163.1 | - | 4 | 505 | 6513848 |