Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ewsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283061.1 | NP_001269990.1 | 3, 4 | 177 | ||
NM_001283062.1 | NP_001269991.1 | 3, 4 | 177 | |||
NM_001283063.1 | NP_001269992.1 | 3, 4 | 177 | |||
NM_007968.3 | NP_031994.2 | 3, 4 | 177 | |||
GenBank mRNA | AK019460.1 | - | 3, 4 | 126 | ||
AK046434.1 | - | 3, 4 | 186 | |||
AK049743.1 | - | 3, 4 | 157 | |||
AK146791.1 | - | 3, 4 | 153 | |||
AK147103.1 | - | 3, 4 | 144 | |||
AK147909.1 | - | 3, 4 | 134 | |||
AK150345.1 | - | 3, 4 | 135 | |||
AK151625.1 | - | 3, 4 | 148 | |||
AK153276.1 | - | 3, 4 | 135 | |||
BC068226.1 | - | 3, 4 | 144 | 5707017 | ||
BY272748.1 | - | 3, 4 | 178 | |||
X79233.1 | - | 3, 4 | 166 |